Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184895.1 | NP_001171824.1 | 1 - 2 | 421 | 99 | |
GenBank mRNA | AK393586.1 | - | 1 - 2 | 128 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |