Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184895.1 | NP_001171824.1 | 3 - 4 | 612 | 57 | |
GenBank mRNA | AK235245.1 | - | 3 - 4 | 314 | 57 | |
AK350347.1 | - | 3 - 4 | 313 | 57 | ||
AK392413.1 | - | 3 - 4 | 317 | 57 | ||
AK393586.1 | - | 3 - 4 | 319 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |