Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185066.1 | NP_001171995.1 | 2 - 3 | 1597 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |