Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM204A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185171.1 | NP_001172100.1 | 8 - 9 | 895 | 68 | |
XM_005671491.3 | XP_005671548.1 | 731 | 68 | |||
XM_005671493.3 | XP_005671550.1 | 647 | 68 | |||
XM_005671494.3 | XP_005671551.1 | 676 | 68 | |||
XM_005671498.3 | XP_005671555.1 | 609 | 68 | |||
XM_021071868.1 | XP_020927527.1 | 819 | 68 | |||
GenBank mRNA | AK234444.1 | - | 8 - 9 | 893 | 68 | |
AK238763.1 | - | 6 - 7 | 605 | 68 | ||
AK351475.1 | - | 6 - 7 | 596 | 68 | ||
AY609511.1 | - | 7 - 8 | 707 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |