Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190159.1 | NP_001177088.1 | 6 - 7 | 941 | 67 | |
XM_005672959.3 | XP_005673016.1 | 986 | 67 | |||
XM_005672960.3 | XP_005673017.1 | 857 | 67 | |||
GenBank mRNA | AK239658.1 | - | 6 - 7 | 941 | 67 | |
AK347559.1 | - | 6 - 7 | 883 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |