Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190175.1 | NP_001177104.1 | 7 - 8 | 897 | 86 | |
GenBank mRNA | AK232518.1 | - | 7 - 8 | 897 | 86 | |
AK237861.1 | - | 7 - 8 | 1298 | 86 | ||
AK346919.1 | - | 7 - 8 | 825 | 86 | ||
AK389542.1 | - | 7 - 8 | 1051 | 86 | ||
AK392778.1 | - | 7 - 8 | 818 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |