Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190181.1 | NP_001177110.1 | 2 - 3 | 351 | 100 | |
XM_021098029.1 | XP_020953688.1 | 344 | 100 | |||
GenBank mRNA | AK231349.1 | - | 2 - 3 | 270 | 100 | |
AK235777.1 | - | 2 - 3 | 184 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |