Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTCAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190186.1 | NP_001177115.1 | 2 - 3 | 244 | 85 | |
GenBank mRNA | AK235351.1 | - | 2 - 3 | 248 | 85 | |
AK238420.1 | - | 2 - 3 | 188 | 85 | ||
AK400712.1 | - | 2 - 3 | 257 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |