Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190189.1 | NP_001177118.1 | 7 - 8 | 867 | 73 | |
XM_021071932.1 | XP_020927591.1 | 1159 | 73 | |||
GenBank mRNA | AK234877.1 | - | 7 - 8 | 867 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |