Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC42SE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190193.1 | NP_001177122.1 | 1 - 2 | 66 | 90 | |
GenBank mRNA | AK239716.1 | - | 1 - 2 | 61 | 90 | |
BW960573.1 | - | 1 - 2 | 66 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |