Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC42SE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190193.1 | NP_001177122.1 | 2 - 3 | 375 | 53 | |
XM_005663461.3 | XP_005663518.1 | 456 | 53 | |||
XM_021088571.1 | XP_020944230.1 | 306 | 53 | |||
GenBank mRNA | AK239716.1 | - | 2 - 3 | 370 | 53 | |
BW960573.1 | - | 2 - 3 | 375 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |