Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15H4orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190210.1 | NP_001177139.1 | 3 - 4 | 533 | 76 | |
XM_005671711.2 | XP_005671768.1 | 666 | 76 | |||
XM_005671712.2 | XP_005671769.1 | 599 | 76 | |||
XM_005671713.3 | XP_005671770.1 | 505 | 76 | |||
XM_005671714.3 | XP_005671771.1 | 1320 | 76 | |||
XM_005671715.2 | XP_005671772.2 | 604 | 76 | |||
XM_013984044.2 | XP_013839498.1 | 629 | 76 | |||
XM_013984045.2 | XP_013839499.1 | 499 | 76 | |||
XM_021074525.1 | XP_020930184.1 | 624 | 76 | |||
GenBank mRNA | AK239144.1 | - | 3 - 4 | 530 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |