Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190225.1 | NP_001177154.1 | 3 - 4 | 382 | 85 | |
XM_013983316.1 | XP_013838770.1 | 422 | 85 | |||
GenBank mRNA | AK232006.1 | - | 3 - 4 | 315 | 85 | |
AK237726.1 | - | 4 - 5 | 354 | 85 | ||
AK238981.1 | - | 4 - 5 | 408 | 85 | ||
AK348720.1 | - | 3 - 4 | 316 | 85 | ||
AK389224.1 | - | 3 - 4 | 377 | 85 | ||
AK391121.1 | - | 3 - 4 | 316 | 85 | ||
AK400580.1 | - | 3 - 4 | 349 | 85 | ||
AY610324.1 | - | 3 - 4 | 276 | 85 | ||
BW971208.1 | - | 3 - 4 | 344 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |