Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPACA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190247.1 | NP_001177176.1 | 5 - 6 | 610 | 108 | |
GenBank mRNA | GU475011.1 | - | 5 - 6 | 610 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |