Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190282.1 | NP_001177211.1 | 3 - 4 | 571 | 88 | |
GenBank mRNA | AK236228.1 | - | 3 - 4 | 595 | 88 | |
AY610153.1 | - | 3 - 4 | 647 | 88 | ||
HM046520.1 | - | 3 - 4 | 571 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Inventoried Probe spans exons |