Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190286.1 | NP_001177215.1 | 5 - 6 | 745 | 70 | |
GenBank mRNA | HM046504.1 | - | 5 - 6 | 745 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |