Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190296.1 | NP_001177225.1 | 2 - 3 | 241 | 61 | |
XM_005656860.3 | XP_005656917.1 | 310 | 61 | |||
GenBank mRNA | HM046503.1 | - | 2 - 3 | 241 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |