Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348539.1 | NP_001335468.1 | 5 - 6 | 664 | 97 | |
XM_021077666.1 | XP_020933325.1 | 1873 | 97 | |||
GenBank mRNA | HM138511.1 | - | 5 - 6 | 664 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons |