Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194972.1 | NP_001181901.1 | 11 - 12 | 1375 | 87 | |
XM_021093284.1 | XP_020948943.1 | 4352 | 87 | |||
XM_021093286.1 | XP_020948945.1 | 1573 | 87 | |||
XM_021093287.1 | XP_020948946.1 | 2798 | 87 | |||
GenBank mRNA | GQ329717.1 | - | 11 - 12 | 1375 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |