Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194976.1 | NP_001181905.1 | 11 - 12 | 1391 | 87 | |
XM_005665209.3 | XP_005665266.1 | 1512 | 87 | |||
XM_021093293.1 | XP_020948952.1 | 1102 | 87 | |||
GenBank mRNA | AK344581.1 | - | 10 - 11 | 1291 | 87 | |
GQ329714.1 | - | 11 - 12 | 1391 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |