Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195118.1 | NP_001182047.1 | 4 - 5 | 484 | 81 | |
XM_005672915.3 | XP_005672972.1 | 821 | 81 | |||
GenBank mRNA | GU576972.1 | - | 4 - 5 | 484 | 81 | |
GU576973.1 | - | 4 - 5 | 366 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |