Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195121.1 | NP_001182050.1 | 7 - 8 | 846 | 59 | |
GenBank mRNA | AK350931.1 | - | 7 - 8 | 845 | 59 | |
HM627270.1 | - | 7 - 8 | 846 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |