Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCORL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195345.1 | NP_001182274.1 | 3 - 4 | 300 | 95 | |
XM_021100293.1 | XP_020955952.1 | 1305 | 95 | |||
XM_021100294.1 | XP_020955953.1 | 535 | 95 | |||
XM_021100295.1 | XP_020955954.1 | 2718 | 95 | |||
XM_021100296.1 | XP_020955955.1 | 373 | 95 | |||
XM_021100297.1 | XP_020955956.1 | 303 | 95 | |||
XM_021100298.1 | XP_020955957.1 | 2753 | 95 | |||
XM_021100299.1 | XP_020955958.1 | 195 | 95 | |||
XM_021100301.1 | XP_020955960.1 | 591 | 95 | |||
XM_021100302.1 | XP_020955961.1 | 337 | 95 | |||
XM_021100303.1 | XP_020955962.1 | 484 | 95 | |||
GenBank mRNA | AK231042.1 | - | 4 - 5 | 408 | 95 | |
GU373703.1 | - | 3 - 4 | 300 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |