Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195352.1 | NP_001182281.1 | 2 - 3 | 140 | 69 | |
XM_005660938.3 | XP_005660995.1 | 540 | 69 | |||
XM_005660939.3 | XP_005660996.1 | 291 | 69 | |||
XM_005660940.2 | XP_005660997.1 | 304 | 69 | |||
GenBank mRNA | AK350068.1 | - | 2 - 3 | 301 | 69 | |
AY610386.1 | - | 2 - 3 | 111 | 69 | ||
GU373711.1 | - | 2 - 3 | 140 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |