Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195359.1 | NP_001182288.1 | 4 - 5 | 719 | 104 | |
XM_005668865.3 | XP_005668922.1 | 1203 | 104 | |||
XM_005668866.3 | XP_005668923.1 | 1204 | 104 | |||
XM_005668867.3 | XP_005668924.1 | 729 | 104 | |||
GenBank mRNA | AK237413.1 | - | 4 - 5 | 733 | 104 | |
AK348542.1 | - | 4 - 5 | 671 | 104 | ||
GU373686.1 | - | 4 - 5 | 719 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |