Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195360.1 | NP_001182289.1 | 1 - 2 | 97 | 59 | |
XM_005668615.3 | XP_005668672.1 | 833 | 59 | |||
GenBank mRNA | GU373685.1 | - | 1 - 2 | 97 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |