Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195363.2 | NP_001182292.1 | 1 - 2 | 118 | 100 | |
GenBank mRNA | AK343386.1 | - | 1 - 2 | 117 | 100 | |
AK348480.1 | - | 1 - 2 | 115 | 100 | ||
GU373669.1 | - | 1 - 2 | 117 | 100 | ||
HX212195.1 | - | 1 - 2 | 119 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |