Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDR16C5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195368.1 | NP_001182297.1 | 2 - 3 | 462 | 103 | |
XM_013996752.2 | XP_013852206.1 | 741 | 103 | |||
GenBank mRNA | AK344212.1 | - | 3 - 4 | 562 | 103 | |
GU373688.1 | - | 2 - 3 | 462 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |