Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss04248083_s1
See other ZBTB38 GE Assays ›
Gene Symbol:
ZBTB38
Gene Name:
zinc finger and BTB domain containing 38
Gene Aliases:
-
Chromosome Location:
Chr.13: 82053887 - 82195321 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss04248083_s1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ZBTB38
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001195373.1
NP_001182302.1
1
3451
65
XM_005669895.3
XP_005669952.1
3704
65
XM_005669896.3
XP_005669953.1
8043
65
XM_005669897.3
XP_005669954.1
5919
65
XM_005669898.3
XP_005669955.1
3686
65
XM_005669899.3
XP_005669956.1
4578
65
XM_013982074.2
XP_013837528.1
4108
65
XM_013982075.2
XP_013837529.1
5203
65
XM_013982078.2
XP_013837532.1
3936
65
XM_021068345.1
XP_020924004.1
4500
65
XM_021068346.1
XP_020924005.1
4467
65
XM_021068347.1
XP_020924006.1
4142
65
XM_021068348.1
XP_020924007.1
4701
65
XM_021068349.1
XP_020924008.1
5966
65
XM_021068350.1
XP_020924009.1
4459
65
XM_021068351.1
XP_020924010.1
4277
65
XM_021068353.1
XP_020924012.1
8401
65
XM_021068354.1
XP_020924013.1
5647
65
XM_021068355.1
XP_020924014.1
5561
65
XM_021068356.1
XP_020924015.1
4105
65
XM_021068357.1
XP_020924016.1
5289
65
XM_021068358.1
XP_020924017.1
5539
65
XM_021068359.1
XP_020924018.1
3857
65
XM_021068360.1
XP_020924019.1
5051
65
XM_021068361.1
XP_020924020.1
4067
65
GenBank mRNA
GU373689.1
-
1
3451
65
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
3' Most
Detect Genomic DNA
Recommended Assay
Sus scrofa
Both primers and probe map within a single exon
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0