Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMYA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197307.1 | NP_001184236.1 | 2 - 3 | 10741 | 102 | |
XM_021077698.1 | XP_020933357.1 | 10781 | 102 | |||
GenBank mRNA | FJ208850.1 | - | 2 - 3 | 10741 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |