Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204382.1 | NP_001191311.1 | 7 - 8 | 776 | 61 | |
XM_021090906.1 | XP_020946565.1 | 1010 | 61 | |||
XM_021090909.1 | XP_020946568.1 | 999 | 61 | |||
XM_021090910.1 | XP_020946569.1 | 740 | 61 | |||
GenBank mRNA | AK350790.1 | - | 7 - 8 | 776 | 61 | |
AK391116.1 | - | 7 - 8 | 932 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |