Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H1FOO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205063.1 | NP_001191992.1 | 3 - 4 | 777 | 73 | |
GenBank mRNA | HQ915640.1 | - | 3 - 4 | 777 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |