Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142669.1 | NP_001136141.1 | 2 | 245 | 109 | |
XM_021076972.1 | XP_020932631.1 | 596 | 109 | |||
XM_021076976.1 | XP_020932635.1 | 596 | 109 | |||
XM_021076979.1 | XP_020932638.1 | 596 | 109 | |||
XM_021076983.1 | XP_020932642.1 | 596 | 109 | |||
XM_021076989.1 | XP_020932648.1 | 596 | 109 | |||
XM_021076995.1 | XP_020932654.1 | 596 | 109 | |||
XM_021077005.1 | XP_020932664.1 | 596 | 109 | |||
XM_021077010.1 | XP_020932669.1 | 246 | 109 | |||
XM_021077020.1 | XP_020932679.1 | 316 | 109 | |||
XM_021077026.1 | XP_020932685.1 | 253 | 109 | |||
XM_021077034.1 | XP_020932693.1 | 595 | 109 | |||
XM_021077040.1 | XP_020932699.1 | 198 | 109 | |||
XM_021077046.1 | XP_020932705.1 | 1544 | 109 | |||
XM_021077049.1 | XP_020932708.1 | 219 | 109 | |||
XM_021077053.1 | XP_020932712.1 | 172 | 109 | |||
GenBank mRNA | AK396842.1 | - | 2 | 513 | 109 | |
EU286536.1 | - | 2 | 245 | 109 | ||
EU286537.1 | - | 2 | 245 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |