Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BZW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244493.1 | NP_001231422.1 | 6 - 7 | 632 | 152 | |
XM_021074514.1 | XP_020930173.1 | 958 | 152 | |||
GenBank mRNA | AK239661.1 | - | 6 - 7 | 638 | 152 | |
AK343867.1 | - | 6 - 7 | 683 | 152 | ||
AK343897.1 | - | 4 - 5 | 478 | 152 | ||
AK345146.1 | - | 1 - 2 | 84 | 152 | ||
AY609907.1 | - | 6 - 7 | 636 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |