Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160084.1 | NP_001153556.1 | 3 - 4 | 496 | 73 | |
GenBank mRNA | FJ875104.1 | - | 3 - 4 | 496 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |