Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166045.1 | NP_001159517.1 | 4 - 5 | 993 | 87 | |
GenBank mRNA | GQ478346.1 | - | 4 - 5 | 993 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |