Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDIPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167642.1 | NP_001161114.1 | 1 - 2 | 47 | 91 | |
XM_005655088.3 | XP_005655145.1 | 384 | 91 | |||
XM_021085630.1 | XP_020941289.1 | 438 | 91 | |||
GenBank mRNA | AB529449.1 | - | 1 - 2 | 47 | 91 | |
AK236165.1 | - | 1 - 2 | 383 | 91 | ||
AK240557.1 | - | 1 - 2 | 429 | 91 | ||
AK351493.1 | - | 1 - 2 | 390 | 91 | ||
AK393252.1 | - | 1 - 2 | 390 | 91 | ||
AK399176.1 | - | 1 - 2 | 392 | 91 | ||
AK400901.1 | - | 1 - 2 | 422 | 91 | ||
GU144288.1 | - | 1 - 2 | 358 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |