Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPACA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177929.1 | NP_001171400.1 | 1 | 219 | 74 | |
GenBank mRNA | GU474992.1 | - | 1 | 219 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |