Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205403.1 | NP_001192332.1 | 2 - 3 | 335 | 106 | |
GenBank mRNA | AK240439.1 | - | 2 - 3 | 327 | 106 | |
AK352168.1 | - | 2 - 3 | 308 | 106 | ||
CJ016107.1 | - | 2 - 3 | 335 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |