Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244921.1 | NP_001231850.1 | 6 - 7 | 755 | 85 | |
XM_005667490.3 | XP_005667547.1 | 786 | 85 | |||
XM_013979554.2 | XP_013835008.1 | 745 | 85 | |||
GenBank mRNA | AK232917.1 | - | 4 - 5 | 377 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |