Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244969.1 | NP_001231898.1 | 6 - 7 | 984 | 77 | |
XM_021079468.1 | XP_020935127.1 | 77 | ||||
GenBank mRNA | AK235534.1 | - | 6 - 7 | 77 | ||
AK346598.1 | - | 6 - 7 | 984 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |