Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244975.1 | NP_001231904.1 | 2 - 3 | 190 | 65 | |
NM_001244976.1 | NP_001231905.1 | 2 - 3 | 165 | 65 | ||
NM_001244977.1 | NP_001231906.1 | 2 - 3 | 165 | 65 | ||
XM_005661298.3 | XP_005661355.1 | 249 | 65 | |||
XM_005661299.3 | XP_005661356.1 | 249 | 65 | |||
GenBank mRNA | AK231677.1 | - | 2 - 3 | 165 | 65 | |
AK234487.1 | - | 2 - 3 | 190 | 65 | ||
AK237847.1 | - | 2 - 3 | 165 | 65 | ||
AK351024.1 | - | 2 - 3 | 165 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |