Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244987.1 | NP_001231916.1 | 3 - 4 | 531 | 61 | |
GenBank mRNA | AK234481.1 | - | 3 - 4 | 531 | 61 | |
AK346168.1 | - | 3 - 4 | 465 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |