Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245006.1 | NP_001231935.1 | 6 - 7 | 756 | 66 | |
XM_005664536.3 | XP_005664593.2 | 751 | 66 | |||
XM_005664537.3 | XP_005664594.2 | 666 | 66 | |||
GenBank mRNA | AK237080.1 | - | 6 - 7 | 756 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |