Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245012.1 | NP_001231941.1 | 4 - 5 | 696 | 97 | |
XM_005672032.3 | XP_005672089.1 | 873 | 97 | |||
XM_005672033.3 | XP_005672090.1 | 1108 | 97 | |||
XM_021074607.1 | XP_020930266.1 | 2881 | 97 | |||
GenBank mRNA | AK232018.1 | - | 4 - 5 | 696 | 97 | |
AK351036.1 | - | 3 - 4 | 517 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |