Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246248.1 | NP_001233177.1 | 6 - 7 | 1706 | 70 | |
GenBank mRNA | AK397678.1 | - | 6 - 7 | 1406 | 70 | |
HQ630069.1 | - | 6 - 7 | 1706 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |