Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PQBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246668.1 | NP_001233597.1 | 1 - 2 | 85 | 68 | |
XM_005673594.3 | XP_005673651.1 | 229 | 68 | |||
XM_013986114.1 | XP_013841568.1 | 495 | 68 | |||
GenBank mRNA | AK231983.1 | - | 2 - 3 | 221 | 68 | |
AK238508.1 | - | 2 - 3 | 500 | 68 | ||
AK351316.1 | - | 2 - 3 | 498 | 68 | ||
AK352148.1 | - | 2 - 3 | 221 | 68 | ||
AK397584.1 | - | 2 - 3 | 224 | 68 | ||
AY609811.1 | - | 129 | 68 | |||
JF750401.1 | - | 1 - 2 | 85 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |