Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206443.1 | NP_001193372.1 | 2 - 3 | 1354 | 91 | |
NM_001206445.1 | NP_001193374.1 | 2 - 3 | 1354 | 91 | ||
NM_001206446.1 | NP_001193375.1 | 2 - 3 | 1354 | 91 | ||
XM_013981369.2 | XP_013836823.1 | 1367 | 91 | |||
XM_021068433.1 | XP_020924092.1 | 601 | 91 | |||
XM_021068434.1 | XP_020924093.1 | 1335 | 91 | |||
GenBank mRNA | AK233563.1 | - | 2 - 3 | 1353 | 91 | |
AK345156.1 | - | 2 - 3 | 1354 | 91 | ||
AK394746.1 | - | 1 - 2 | 118 | 91 | ||
CK459948.1 | - | 3 - 4 | 631 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |