Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243056.1 | NP_001229985.1 | 7 - 8 | 1165 | 68 | |
XM_021093328.1 | XP_020948987.1 | 1272 | 68 | |||
XM_021093330.1 | XP_020948989.1 | 1371 | 68 | |||
XM_021093331.1 | XP_020948990.1 | 1117 | 68 | |||
XM_021093332.1 | XP_020948991.1 | 1093 | 68 | |||
XM_021093333.1 | XP_020948992.1 | 1567 | 68 | |||
XM_021093334.1 | XP_020948993.1 | 1289 | 68 | |||
XM_021093335.1 | XP_020948994.1 | 1147 | 68 | |||
XM_021093336.1 | XP_020948995.1 | 2703 | 68 | |||
XM_021093337.1 | XP_020948996.1 | 1326 | 68 | |||
XM_021093338.1 | XP_020948997.1 | 1241 | 68 | |||
GenBank mRNA | AK348726.1 | - | 7 - 8 | 1165 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Sus scrofa Probe spans exons |