Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243260.1 | NP_001230189.1 | 2 - 3 | 556 | 82 | |
GenBank mRNA | AK239290.1 | - | 2 - 3 | 513 | 82 | |
AK392669.1 | - | 2 - 3 | 510 | 82 | ||
FS661335.1 | - | 2 - 3 | 515 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |